Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-6R (soluble) Recombinant Protein, PeproTech®

Catalog No. 20006RC500
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
5 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20006RC500 500 μg
20006RC100 100 μg
20006RC1MG 1 mg
20006RC20UG 20 μg
20006RC50UG 50 μg
20006RC5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 20006RC500 Supplier Gibco™ Supplier No. 20006RC500UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sIL-6 Receptor alpha is a 37.9 kDa glycoprotein corresponding to 339 amino acid residues of the extracellular domain of IL-6Ralpha. As a result of glycosylation, Recombinant Human sIL-6 Receptor alpha migrates with an apparent molecular mass of approximately 57-70 kDa by SDS-PAGE gel, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08887
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3570
Molecular Weight (g/mol) 37.9 kDa
Name Human IL-6 R (soluble)
Quantity 500 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6 R; IL-6 R 1; IL-6 R subunit alpha; IL6R1; IL-6 R-1; Il6ra; IL-6 RA; IL-6 R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991
Common Name IL-6 Receptor (CD126)
Gene Symbol IL6R
Biological Activity Determined by its ability to intensify the IL-6 induced growth inhibition of mouse M1 cells. The expected ED50 is ≤ 5.0 ng/ml, in the presence of 20 ng/ml of rhIL-6.
Conjugate Unconjugated
Recombinant Recombinant
Sequence LAPRRCPAQE VARGVLTSLP GDSVTLTCPG VEPEDNATVH WVLRKPAAGS HPSRWAGMGR RLLLRSVQLH DSGNYSCYRA GRPAGTVHLL VDVPPEEPQL SCFRKSPLSN VVCEWGPRST PSLTTKAVLL VRKFQNSPAE DFQEPCQYSQ ESQKFSCQLA VPEGDSSFYI VSMCVASSVG SKFSKTQTFQ GCGILQPDPP ANITVTAVAR NPRWLSVTWQ DPHSWNSSFY RLRFELRYRA ERSKTFTTWM VKDLQHHCVI HDAWSGLRHV VQLRAQEEFG QGEWSEWSPE AMGTPWTESR SPPAENEVST PMQALTTNKD DDNILFRDSA NATSLPVQD
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.