Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-7 Recombinant Protein, PeproTech®

Catalog No. 2000701M
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-007-01M 2 x 500 μg
20007100UG 100 μg
200-07-10UG 10 μg
20007250UG 250 μg
200-07-2UG 2 μg
20007500UG 500 μg
200-07-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 20-007-01M Supplier Gibco™ Supplier No. 2000701M
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

200-07-1MG will be provided as 2 x 500 μg (200-07-500UG). Recombinant Human IL-7 is a 17.5 kDa protein containing 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13232, P16871
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3574, 3575
Molecular Weight (g/mol) 17.5 kDa
Name Human IL-7
Purification Method Purified
Quantity 2 x 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7
Common Name IL-7
Gene Symbol Il7, IL7r
Biological Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine 2E8 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH
Content And Storage -20°C
Activity The ED50 was determined by the dose-dependent stimulation of the proliferation of murine 2E8 cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.