Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-8/CXCL8 Quantikine HS ELISA Kit

Catalog No. HS800
Change view
Click to view available options
Quantity:
1 Pack (6 Plates)
One 96-well plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HS800 One 96-well plate
SS800 1 Pack (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. HS800 Supplier R&D Systems™ Supplier No. HS800
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human IL-8/CXCL8 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-8/CXCL8 Quantikine HS ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL8/IL-8 in serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).

Specifications

Assay Range 1.00 - 64 pg/mL
Assay Sensitivity 0.4 pg/mL
Conjugate HRP
Sample Type Plasma (EDTA), Heparin Plasma, Serum, Serum
Specificity Natural and Recombinant Human IL-8
For Use With (Application) ELISA
Gene ID (Entrez) 3576
Quantity One 96-well plate
Sample Volume Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym 3-10 C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1 TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1 NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10 C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.