Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL12A (P29459, 503 amino acids) Partial Recombinant Protein expressed in CHO cells

Catalog No. 89948620 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-620 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-620 Supplier Abnova™ Supplier No. P3997

Used for Func, SDS-PAGE

This gene encodes a subunit of a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. The cytokine is a disulfide-linked heterodimer composed of the 35-kD subunit encoded by this gene, and a 40-kD subunit that is a member of the cytokine receptor family. This cytokine is required for the T-cell-independent induction of interferon (IFN)-gamma, and is important for the differentiation of both Th1 and Th2 cells. The responses of lymphocytes to this cytokine are mediated by the activator of transcription protein STAT4. Nitric oxide synthase 2A (NOS2A/NOS2) is found to be required for the signaling process of this cytokine in innate immunity. [provided by RefSeq]

Specifications

Accession Number P29459
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 3592
Molecular Weight (g/mol) 75kDa
Name IL12A (Human) Recombinant Protein
Preparation Method Mammalian cell (CHO) expression system
Quantity 10 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with PBS, store at 4°C for 1 week or -20°C for longer. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias CLMF/IL-12A/NFSK/NKSF1/P35
Common Name IL12A
Gene Symbol IL12A
Biological Activity The ED50 was determined by the induction of IFN-g production from murine splenocytes co-stimulated with IL-12 is < 0.1ng/mL, corresponding to a specific activity of > 1 x 107 units/mg.
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (CHO) expression system
Form Lyophilized
Purity or Quality Grade ≥98% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.