Learn More
Sino Biological Human IL-35 (IL12A & EBI3 Heterodimer) Protein 500ug
Supplier:  Sino Biological CT103H2303H500
Encompass_Preferred
A DNA sequence encoding the p35 subunit of human IL12, termed as IL12A (P29459-1) (Met1-Ser219) was fused with a C-terminal Fc region of human IgG1 tag followed by a FLAG tag, constructed the plasmid 1; A DNA sequence encoding the human IL27B (NP_005746.2) (Met1-Lys229) was fused with a C-terminal Fc region of human IgG1 tag followed by a polyhistidine tag, constructed the plasmid 2. The two plasmids were co-expressed and the heterodimer was purified.
Catalog No. 50-255-2595
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.