Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human IL-12/IL-23 p40 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP182421 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP182421 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP182421 Supplier Novus Biologicals™ Supplier No. NBP182421

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human IL12B ELISA Kit (Colorimetric) measures human IL12B in cell culture supernatant, serum, plasma (citrate, heparin).

Specifications

Assay Range 31.3 - 2000pg/mL
Assay Sensitivity <10pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (Citrate, Heparin), Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3593
Gene Symbol IL12B
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human IL-12/IL-23 p40, Biotin-Conjugate anti-human IL-12/IL-23 p40 monoclonal antibody, Streptavidin-HRP, Human IL-12/IL-23 p40 Standard lyophilized, 4ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration facto
Target IL12B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.