Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL13 (P35225) Partial Recombinant Protein

Catalog No. 89948641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-641 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-641 Supplier Abnova™ Supplier No. P6089

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for Func, SDS-PAGE

This gene encodes an immunoregulatory cytokine produced primarily by activated Th2 cells. This cytokine is involved in several stages of B-cell maturation and differentiation. It up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. This cytokine down-regulates macrophage activity, thereby inhibits the production of pro-inflammatory cytokines and chemokines. This cytokine is found to be critical to the pathogenesis of allergen-induced asthma but operates through mechanisms independent of IgE and eosinophils. This gene, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with this gene particularly close to IL4. [provided by RefSeq]

Sequence: MSPGPVPPSTALRELIEELVNITQNQKAPLCNGSMVWSINLTAGMYCAALESLINVSGCSAIEKTQRMLSGFCPHKVSAGQFSSLHVRDTKIEVAQFVKDLLLHLKKLFREGQFN

Specifications

Accession Number P35225
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 3596
Name IL13 (Human) Recombinant Protein
Preparation Method E. coli expression system
Quantity 10 μg
Immunogen MSPGPVPPSTALRELIEELVNITQNQKAPLCNGSMVWSINLTAGMYCAALESLINVSGCSAIEKTQRMLSGFCPHKVSAGQFSSLHVRDTKIEVAQFVKDLLLHLKKLFREGQFN
Storage Requirements Store at -20°C. Reconstitute in water to a concentration of 0.5-1.0 mg/mL. Do not vortex. Note: Allow the reconstituted vial to sit at 4°C for more than 2 hours before use. For extended storage, it is recommended to be further diluted in buffer containing carrier protein (e.g. 0.1% BSA) and stored in aliquots at -20°C to -80°C.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (<1 EU/μg)
Gene Alias ALRH/BHR1/IL-13/MGC116786/MGC116788/MGC116789/P600
Common Name IL13
Gene Symbol IL13
Biological Activity This IL-13 analog, with a substitution of Q for R at position 112, measured by the in vitro dose dependent activation of STAT6 and IL-13 dependent gene induction in transfected A201.1 cells. This analog has also been shown to exhibit increased in vivo activity compared to wild type IL-13, as measured by the induction of airway hyper-responsiveness.
Cross Reactivity Human ;Human
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System E. coli expression system
Form Lyophilized
Purity or Quality Grade 0.98
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.