Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL16 Full-length ORF (AAH40272.1, 1 a.a. - 454 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89948670 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-670 10 ug
89-948-671 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-948-670 Supplier Abnova™ Supplier No. H00003603P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of this gene undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MESHSRAGKSRKSAKFRSISRSLMLCNAKTSDDGSSPDEKYPDPFEISLAQGKEGIFHSSVQLADTSEAGPSSVPDLALASEAAQLQAAGNDRGKTCRRIFFMKESSTASSREKPGKLEAQSSNFLFPKACHQRARSNSTSVNPYCTREIDFPMTKKSAAPTDRQPYSLCSNRKSLSQQLDCPAGKAAGTSRPTRSLSTAQLVQPSGGLQASVISNIVLMKGQAKGLGFSIVGGKDSIYGPIGIYVKTIFAGGAAAADGRLQEGDEILELNGESMAGLTHQDALQKFKQAKKGLLTLTVRTRLTAPPSLCSHLSPPLCRSLSSSTCITKDSSSFALESPSAPISTAKPNYRIMVEVSLQKEAGVGLGIGLCSVPYFQCISGIFVHTLSPGSVAHLDGRLRCGDEIVEISDSPVHCLTLNEVYTILSHCDPGPVSIIVSRHPDPQKEEMEAQEVK

Specifications

Accession Number AAH40272.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3603
Molecular Weight (g/mol) 74.8kDa
Name IL16 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MESHSRAGKSRKSAKFRSISRSLMLCNAKTSDDGSSPDEKYPDPFEISLAQGKEGIFHSSVQLADTSEAGPSSVPDLALASEAAQLQAAGNDRGKTCRRIFFMKESSTASSREKPGKLEAQSSNFLFPKACHQRARSNSTSVNPYCTREIDFPMTKKSAAPTDRQPYSLCSNRKSLSQQLDCPAGKAAGTSRPTRSLSTAQLVQPSGGLQASVISNIVLMKGQAKGLGFSIVGGKDSIYGPIGIYVKTIFAGGAAAADGRLQEGDEILELNGESMAGLTHQDALQKFKQAKKGLLTLTVRTRLTAPPSLCSHLSPPLCRSLSSSTCITKDSSSFALESPSAPISTAKPNYRIMVEVSLQKEAGVGLGIGLCSVPYFQCISGIFVHTLSPGSVAHLDGRLRCGDEIVEISDSPVHCLTLNEVYTILSHCDPGPVSIIVSRHPDPQKEEMEAQEVK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ16806/FLJ42735/FLJ44234/HsT19289/IL-16/LCF/prIL-16
Common Name IL16
Gene Symbol IL16
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.