Learn More
Sino Biological Human IL18R1 MAb 20UL 11102-R011-20
Supplier:  Sino Biological 11102R01120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL18R1 / CD218a (rh IL18R1; Catalog#11102-H08H; NP_003846.1; Met 1-Arg 329).
Catalog No. 50-240-8014
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.