Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL2 (133 a.a.) Full-length Recombinant Protein with His tag expressed in Barley grain

Catalog No. 89948792 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-792 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-792 Supplier Abnova™ Supplier No. P5822

Used for Func, SDS-PAGE, WB-Re

The protein encoded by this gene is a secreted cytokine that is important for the proliferation of T and B lymphocytes. The receptor of this cytokine is a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL4) and interleukin 7 (IL7). The expression of this gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests an essential role of this gene in the immune response to antigenic stimuli. [provided by RefSeq]

Specifications

Accession Number A14844
Concentration 100μg/mL
For Use With (Application) Functional Assay, SDS-PAGE, Western Blot
Formulation Lyophilized from PBS, pH 7.2
Gene ID (Entrez) 3558
Molecular Weight (g/mol) 22kDa
Name Human IL2 (133 a.a.) Full-length Recombinant Protein with His tag expressed in Barley grain
Purification Method Chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 1 mg
Storage Requirements Store at −20°C on dry atmosphere.After reconstitution with sterile water to a concentration of no less than 100 ug/mL, store at −20°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA).Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.005ng/μg protein (0.05EU/μg) as measured by turbidimetric kinetic assay
Gene Alias IL-2, TCGF, lymphokine
Common Name IL2
Gene Symbol IL2
Biological Activity Bioactivity of recombinant human IL2 is assayed by measuring its dose dependent effect on proliferation of CTLL-2 cells. The ED50 for this effect is typically < 0.25ng/mL, corresponding to a specific activity > 4 x106U/mg.
Species Plants
Recombinant Recombinant
Protein Tag His Tag
Expression System Barley grain (Hordeum vulgare) expression system
Form Lyophilized
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.