Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL2RB (P14784, Ala 27- Asp 239) Partial Recombinant Protein expressed in HEK293 cells

Catalog No. 89948869 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-869 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-869 Supplier Abnova™ Supplier No. P3675
Add to Cart
Add to Cart

Used for SDS-PAGE

The interleukin 2 receptor, which is involved in T cell-mediated immune responses, is present in 3 forms with respect to ability to bind interleukin 2. The low affinity form is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from interleukin 2. The protein encoded by this gene represents the beta subunit and is a type I membrane protein. [provided by RefSeq]

Specifications

Accession Number P14784
For Use With (Application) SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 3560
Molecular Weight (g/mol) 42kDa
Name IL2RB (Human) Recombinant Protein
Preparation Method Mammalian cell (HEK293) expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 5 μg
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 12 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1 EU/μg)
Gene Alias CD122/P70-75
Common Name IL2RB
Gene Symbol IL2RB
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (HEK293) expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.