Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL3 (149 a.a.) Full-lenth Recombinant Protein with His tag expressed in Barley grain (Hordeum vulgare)

Catalog No. 89948875 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-875 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-875 Supplier Abnova™ Supplier No. P5884

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for Func, SDS-PAGE, WB-Re

The protein encoded by this gene is a potent growth promoting cytokine. This cytokine is capable of supporting the proliferation of a broad range of hematopoietic cell types. It is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. This cytokine has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders. [provided by RefSeq]

Specifications

For Use With (Application) Functional Assay, SDS-PAGE, Western Blot
Formulation Lyophilized from PBS, pH 7.2
Gene ID (Entrez) 3562
Molecular Weight (g/mol) 20-23kDa
Name Human IL3 (149 a.a.) Full-lenth Recombinant Protein with His tag expressed in Barley grain (Hordeum vulgare)
Purification Method Chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 1 mg
Storage Requirements Store at −20°C.Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.005ng/μg (0.05EU/μg) as measured by turbidimetric kineticassay.
Gene Alias IL-3, MCGF, MGC79398, MGC79399, MULTI-CSF
Common Name IL3
Gene Symbol IL3
Biological Activity Activity has been measured by cell-based proliferation assay using TF-1 cells. ED50 is 0.18ng/mL.
Species Plants
Recombinant Recombinant
Protein Tag His Tag
Expression System Hordeum vulgare (barley) expression system
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.