Learn More
Sino Biological Human IL3 PAb 100UL 11858-T16-100
Supplier:  Sino Biological 11858T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL3 / IL-3 (rh IL3 / IL-3; Catalog#11858-H08B; NP_000579.2; Met1-Phe152). IL3 / IL-3 specific IgG was purified by Human IL3 / IL-3 affinity chromatography.
Catalog No. 50-241-2604
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.