Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL5RA Full-length ORF (NP_783852.1) Recombinant Protein without tag

Catalog No. 89948931 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-931 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-931 Supplier Abnova™ Supplier No. H00003568G01
Add to Cart
Add to Cart

Used for AP, Func, Screening

The protein encoded by this gene is an interleukin 5 specific subunit of a heterodimeric cytokine receptor. The receptor is comprised of a ligand specific alpha subunit and a signal transducing beta subunit shared by the receptors for interleukin 3 (IL3), colony stimulating factor 2 (CSF2/GM-CSF), and interleukin 5 (IL5). The binding of this protein to IL5 depends on the beta subunit. The beta subunit is activated by the ligand binding, and is required for the biological activities of IL5. This protein has been found to interact with syndecan binding protein (syntenin), which is required for IL5 mediated activation of the transcription factor SOX4. Six alternatively spliced transcript variants encoding three distinct isoforms have been reported. [provided by RefSeq]

Sequence: MIIVAHVLLILLGATEILQADLLPDEKISLLPPVNFTIKVTGLAQVLLQWKPNPDQEQRNVNLEYQVKINAPKEDDYETRITESKCVTILHKGFSASVRTILQNDHSLLASSWASAELHAPPGSPGTSIVNLTCTTNTTEDNYSRLRSYQVSLHCTWLVGTDAPEDTQYFLYYRYGSWTEECQEYSKDTLGRNIACWFPRTFILSKGRDWLAVLVNGSSKHSAIRPFDQLFALHAIDQINPPLNVTAEIEGTRLSIQWEKPVSAFPIHCFDYEVKIHNTRNGYLQIEKLMTNAFISIIDDLSKYDVQVRAAVSSMCREAGLWSEWSQPIYVGFSR

Specifications

Accession Number NP_783852.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 3568
Molecular Weight (g/mol) 38kDa
Name IL5RA (Human) Recombinant Protein
Quantity 10 μg
Immunogen MIIVAHVLLILLGATEILQADLLPDEKISLLPPVNFTIKVTGLAQVLLQWKPNPDQEQRNVNLEYQVKINAPKEDDYETRITESKCVTILHKGFSASVRTILQNDHSLLASSWASAELHAPPGSPGTSIVNLTCTTNTTEDNYSRLRSYQVSLHCTWLVGTDAPEDTQYFLYYRYGSWTEECQEYSKDTLGRNIACWFPRTFILSKGRDWLAVLVNGSSKHSAIRPFDQLFALHAIDQINPPLNVTAEIEGTRLSIQWEKPVSAFPIHCFDYEVKIHNTRNGYLQIEKLMTNAFISIIDDLSKYDVQVRAAVSSMCREAGLWSEWSQPIYVGFSR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CD125/CDw125/HSIL5R3/IL5R/MGC26560
Common Name IL5RA
Gene Symbol IL5RA
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.