Learn More
Sino Biological Recombinant Human IL-7 Protein, GMP grade, HPLC-verified
Supplier:  Sino Biological GMP11821HNAEL25
Encompass_Preferred
A DNA sequence encoding the Human IL7 (NP_000871.1) (Asp26-His177) was expressed with a N-terminal Met.
Catalog No. 50-255-2584