Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ILDR2 (NP_955383) VersaClone cDNA

Catalog No. RDC2240
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2240 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2240 Supplier R&D Systems™ Supplier No. RDC2240
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Human ILDR2 (NP_955383) VersaClone cDNA belongs to the immunoglobulin superfamily.

The Human ILDR2 (NP_955383) VersaClone cDNA clone contains the complete ORF for ILDR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ILDR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Specifications

Gene hILDR2
Accession Number NP_955383
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 1933bp
pH 8.5
Gene ID (Entrez) 387597
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.