Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ILT-2/LIR-1/CD85j ELISA Kit

Catalog No. EH277RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH277RB 96 Tests
EH277RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH277RB Supplier Invitrogen™ Supplier No. EH277RB
Add to Cart
Add to Cart

ELISA

Human ILT-2/LIR-1/CD85j quantitates human ILT-2/LIR-1/CD85j in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ILT-2/LIR-1/CD85j.

This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CD85 antigen-like family member J; CD85j; Ig-like transcript 2; ILT2; ILT-2; Immunoglobulin-like transcript 2; leucocyte Ig-like receptor B1; leukocyte immunoglobulin like receptor B1; leukocyte immunoglobulin-like receptor subfamily B member 1; leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform; leukocyte immunoglobulin-like receptor subfamily B member 3-like; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LILRB1; LIR1; LIR-1; LOC100912456; MIR7; MIR-7; Monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; PIRB; PIR-B
Assay Range 22 to 5,000 pg/mL
Assay Sensitivity 22 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 10859
Gene Symbol LILRB1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CD85j (ILT2)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.