Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ILT1/CD85h APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6364A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6364A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6364A Supplier R&D Systems Supplier No. FAB6364A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT1/CD85h/LILRA2 Monoclonal specifically detects ILT1/CD85h/LILRA2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ILT1/CD85h/LILRA2
Applications Flow Cytometry
Classification Monoclonal
Clone 600007
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8N149
Gene Alias CD85 antigen-like family member H, CD85h, CD85h antigen, ILT1, ILT-1, ILT1CD85H, Leukocyte immunoglobulin-like receptor 7, leukocyte immunoglobulin-like receptor subfamily A member 2, leukocyte immunoglobulin-like receptor subfamily A member 2 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2, LIR7, LIR7LIR-7Immunoglobulin-like transcript 1
Gene Symbols LILRA2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT1/CD85h Pro17-Ser437 Accession # Q8N149
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11027
Test Specificity Detects human ILT1/CD85h in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT2, 3, 4, 5, rhLIR6, or rhLIR8 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.