Learn More
Novus Biologicals™ Human ILT4/CD85d/LILRB2 ELISA Kit (Chemiluminescence)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 31.25 to 2000 pg/mL |
| Assay Sensitivity | 18.75 pg/mL |
| Detection Method | Chemiluminescence |
| Sample Type | Tissue Homogenates, Cell Lysates, Other Biological Fluids |
| Specificity | This kit recognizes HumanI LT4/CD85d/LILRB2 in samples. No significant cross-reactivity or interference between Human LILRB2 and analogues was observed |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 10288 |
| Gene Symbol | LILRB2 |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.