Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Human Immunodeficiency Virus Type 1 (HIV-1) Culture Fluid
SDP

Catalog No. 22157485 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Strain Designations:
BaL
IIIB
Mn
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Strain Designations
22-157-485 IIIB
22-157-578 Mn
22-157-580 BaL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Contains live and infectious organisms.
Catalog No. 22-157-485 Supplier ZeptoMetrix Supplier No. 0801032CF
Add to Cart
Add to Cart
Contains live and infectious organisms.

Infectious virus preparation designed to meet research and product development needs

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Content And Storage -65°C
Form Solution
Media Type Vial
Product Type Human Immunodeficiency Virus Infectious Culture Fluid
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 mL
Strain Designations IIIB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.