Learn More
ZeptoMetrix Human Immunodeficiency Virus Type 1 (HIV-1) Culture Fluid
Shop All ZeptoMetrix Corporation Products

Description
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
Specifications
| Biosafety Level | 2 |
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | -65°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Human Immunodeficiency Virus Infectious Culture Fluid |
| For Use With (Application) | Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
| Quantity | 1 mL |
| Strain Designations | Mn |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.