Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ING3 Full-length ORF (NP_061944.2, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949037 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-037 10 μg
89-949-038 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-949-037 Supplier Abnova™ Supplier No. H00054556P02S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This gene can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. Overexpression of this gene has been shown to inhibit cell growth and induce apoptosis. Allelic loss and reduced expression of this gene were detected in head and neck cancers. Two alternatively spliced transcript variants encoding different isoforms have been observed. [provided by RefSeq]

Sequence: MLYLEDYLEMIEQLPMDLRDRFTEMREMDLQVQNAMDQLEQRVSEFFMNAKKNKPEWREEQMASIKKDYYKALEDADEKVQLANQIYDLVDRHLRKLDQELAKFKMELEADNAGITEILERRSLELDTPSQPVNNHHAHSHTPVEKRKYNPTSHHTTTDHIPEKKFKSEALLSTLTSDASKENTLGCRNNNSTASSNNAYNVNSSQPLGSYNIGSLSSGTGAGAITMAAAQAVQATAQMKEGRRTSSLKASYEAFKNNDFQLGKEFSMARETVGYSSSSALMTTLTQNASSSAADSRSGRKSKNNNKSSSQQSSSSSSSSSLSSCSSSSTVVQEISQQTTVVPESDSNSQVDWTYDPNEPRYCICNQVSYGEMVGCDNQDCPIEWFHYGCVGLTEAPKGKWYCPQCTAAMKRRGSRHK

Specifications

Accession Number NP_061944.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54556
Molecular Weight (g/mol) 73.1kDa
Name ING3 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MLYLEDYLEMIEQLPMDLRDRFTEMREMDLQVQNAMDQLEQRVSEFFMNAKKNKPEWREEQMASIKKDYYKALEDADEKVQLANQIYDLVDRHLRKLDQELAKFKMELEADNAGITEILERRSLELDTPSQPVNNHHAHSHTPVEKRKYNPTSHHTTTDHIPEKKFKSEALLSTLTSDASKENTLGCRNNNSTASSNNAYNVNSSQPLGSYNIGSLSSGTGAGAITMAAAQAVQATAQMKEGRRTSSLKASYEAFKNNDFQLGKEFSMARETVGYSSSSALMTTLTQNASSSAADSRSGRKSKNNNKSSSQQSSSSSSSSSLSSCSSSSTVVQEISQQTTVVPESDSNSQVDWTYDPNEPRYCICNQVSYGEMVGCDNQDCPIEWFHYGCVGLTEAPKGKWYCPQCTAAMKRRGSRHK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias Eaf4/FLJ20089/ING2/p47ING3
Common Name ING3
Gene Symbol ING3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.