Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ING5 Full-length ORF (NP_115705.2, 1 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949046 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-046 10 μg
89-949-047 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-949-046 Supplier Abnova™ Supplier No. H00084289P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This protein can bind TP53 and EP300/p300, a component of the histone acetyl transferase complex, suggesting its involvement in TP53-dependent regulatory pathway. [provided by RefSeq]

Sequence: MATAMYLEHYLDSIENLPCELQRNFQLMRELDQRTEDKKAEIDILAAEYISTVKTLSPDQRVERLQKIQNAYSKCKEYSDDKVQLAMQTYEMVDKHIRRLDADLARFEADLKDKMEGSDFESSGGRGLKKGRGQKEKRGSRGRGRRTSEEDTPKKKKHKGGSEFTDTILSVHPSDVLDMPVDPNEPTYCLCHQVSYGEMIGCDNPDCPIEWFHFACVDLTTKPKGKWFCPRCVQEKRKKK

Specifications

Accession Number NP_115705.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84289
Molecular Weight (g/mol) 54.2kDa
Name ING5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MATAMYLEHYLDSIENLPCELQRNFQLMRELDQRTEDKKAEIDILAAEYISTVKTLSPDQRVERLQKIQNAYSKCKEYSDDKVQLAMQTYEMVDKHIRRLDADLARFEADLKDKMEGSDFESSGGRGLKKGRGQKEKRGSRGRGRRTSEEDTPKKKKHKGGSEFTDTILSVHPSDVLDMPVDPNEPTYCLCHQVSYGEMIGCDNPDCPIEWFHFACVDLTTKPKGKWFCPRCVQEKRKKK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ23842/p28ING5
Common Name ING5
Gene Symbol ING5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.