Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human INHBE Full-length ORF (AAH05161, 21 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949060 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-060 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-949-060 Supplier Abnova™ Supplier No. H00083729P01

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

INHBE is a member of the activin beta family (see INHBA; MIM 147290) that plays a role in pancreatic exocrine cell growth and proliferation (Hashimoto et al., 2006 [PubMed 16426570]).[supplied by OMIM]

Sequence: GTGSVCPSCGGSKLAPQAERALVLELAKQQILDGLHLTSRPRITHPPPQAALTRALRRLQPGSVAPGNGEEVISFATVTDSTSAYSSLLTFHLSTPRSHHLYHARLWLHVLPTLPGTLCLRIFRWGPRRRRQGSRTLLAEHHITNLGWHTLTLPSSGLRGEKSGVLKLQLDCRPLEGNSTVTGQPRRLLDTAGHQQPFLELKIRANEPGAGRARRRTPTCEPATPLCCRRDHYVDFQELGWRDWILQPEGYQLNYCSGQCPPHLAGSPGIAASFHSAVFSLLKANNPWPASTSCCVPTARRPLSLLYLDHNGNVVKTDVPDMVVEACGCS

Specifications

Accession Number AAH05161
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 83729
Molecular Weight (g/mol) 62.04kDa
Name INHBE (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen GTGSVCPSCGGSKLAPQAERALVLELAKQQILDGLHLTSRPRITHPPPQAALTRALRRLQPGSVAPGNGEEVISFATVTDSTSAYSSLLTFHLSTPRSHHLYHARLWLHVLPTLPGTLCLRIFRWGPRRRRQGSRTLLAEHHITNLGWHTLTLPSSGLRGEKSGVLKLQLDCRPLEGNSTVTGQPRRLLDTAGHQQPFLELKIRANEPGAGRARRRTPTCEPATPLCCRRDHYVDFQELGWRDWILQPEGYQLNYCSGQCPPHLAGSPGIAASFHSAVFSLLKANNPWPASTSCCVPTARRPLSLLYLDHNGNVVKTDVPDMVVEACGCS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC4638
Common Name INHBE
Gene Symbol INHBE
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.