Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human INSL5/INSL7 Recombinant Protein, PeproTech®

Catalog No. 130055UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
130-05-5UG 5 μg
13005100UG 100 μg
130-05-1MG 1 mg
13005250UG 250 μg
130-05-25UG 25 μg
13005500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 130-05-5UG Supplier Gibco™ Supplier No. 130055UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human INSL5/INSL7 hybrid is a heterodimeric, 5.2 kDa protein consisting of a 27 amino acid beta-chain linked to a 21 amino acid alpha-chain by two disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The Insulin/Relaxin structural family of proteins consists of at least seven members, each containing an insulin-like heterodimeric structure of two polypeptide chains held together by two inter-chain disulfide bonds. The INSL5/INSL7 hybrid contains the beta-chain of INSL7 (Relaxin-3), linked to the alpha-chain of INSL5. The INSL5/INSL7 hybrid protein can specifically activate the G protein-coupled receptors, GPCR135 and GPCR142, but unlike Relaxin-3, does not induce significant activation of the LGR7 receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WXF3, Q9Y5Q6
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 10022, 117579
Molecular Weight (g/mol) 5.2 kDa
Name Human INSL5/INSL7
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name INSL5/INSL7
Gene Symbol INSL5, RLN3
Conjugate Unconjugated
Recombinant Recombinant
Sequence A-Chain: QDLQTLCCTD GCSMTDLSAL C B-Chain: RAAPYGVRLC GREFIRAVIF TCGGSRW
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.