Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human INSR (NM_000208, 989 a.a. - 1382 a.a.) Partial Recombinant Protein with GST-tag expressed in Sf9 cells

Catalog No. 89949102 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-102 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-949-102 Supplier Abnova™ Supplier No. P4705
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

After removal of the precursor signal peptide, the insulin receptor precursor is post-translationally cleaved into two chains (alpha and beta) that are covalently linked. Binding of insulin to the insulin receptor (INSR) stimulates glucose uptake. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Accession Number NM_000208
Concentration 0.189 ug/uL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 3643
Molecular Weight (g/mol) 70.392kDa
Name INSR (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Purification Method GST affinity chromatography
Quality Control Testing 2 ug/lane SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing
Regulatory Status RUO
Gene Alias CD220/HHF5
Common Name INSR
Gene Symbol INSR
Biological Activity 533pmol/μg x min
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Insect cell (Sf9) expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.