Learn More
Biomatik Corporation Human Insulin Like Protein 5 (INSL5) ELISA Kit
Supplier:  Biomatik Corporation EKN4612696T
Encompass_Preferred
Human Insulin Like Protein 5 (INSL5) ELISA Kit for detecting INSL5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5Q6. Gene ID: 10022. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC2582403
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.