Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Insulin Recombinant Protein

Catalog No. PIRP10908
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP10908 25 mg
PIRP10935 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP10908 Supplier Invitrogen™ Supplier No. RP10908
Add to Cart
Edge
Add to Cart

Recombinant Protein

This is a recombinant protein suitable for use as a control in ELISA and Western blot applications. Lyophilized Insulin is stable at room temperature for 3 weeks. Reconstitute in sterile 0.005N HCl to a concentration of no more than 1mg/mL. Store lyophilized product at -20°C. Upon reconstitution, store at 4°C for 2-7 days or aliquot and store at -20°C. For long term storage add either HSA or BSA to 0.1%. Avoid repeated freeze-thaw cycles.

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01308
Concentration 1 mg/mL
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3630
Molecular Weight (g/mol) 5.8 kDa
Name Human Insulin
Purification Method Purified
Quantity 25 mg
Source E. coli
Immunogen N/A
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulin
Common Name Insulin
Gene Symbol INS
Biological Activity Fully biologically active as compared to WHO reference standard at 28 U/mg
Product Type Protein
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Human Insulin is a two chain, non-glycosylated polypeptide containing 51 amino acids.
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Fully biologically active as compared to WHO reference standard at 28 U/mg.
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.