Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha M/CD11b Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems FAB16991G100
This item is not returnable.
View return policy
Description
CD11b Monoclonal specifically detects CD11b in Human samples. It is validated for Flow Cytometry.Specifications
CD11b | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3684 | |
Human | |
Purified |
Flow Cytometry | |
238446 | |
Flow Cytometry 5 uL/10^6 cells | |
NP_001139280 | |
ITGAM | |
Mouse myeloma cell line NS0-derived recombinant human Integrin alpha M/CD11b Phe17-Asn1105 Accession # NP_001139280 | |
100 Tests | |
Primary | |
Detects human Integrin alpha M/CD11b in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction