Learn More
Bioworld HUMAN INTERFERON-ALPHA 2B (
Supplier:  Bioworld 220603651
Encompass_Preferred
HUMAN INTERFERON-ALPHA 2B (RECOMBINANT): Recombinant Human Interferon-alpha 2b (IFN-a) Source: E.Coli Background: At least 23 different variants of IFN-alpha are known. The individual proteins have molecular masses between 19-26 kDa ...
Catalog No. 50-253-462
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.