Learn More
Creative Biomart Interferon beta-1a
Supplier:  Creative Biomart THP0181
Encompass_Preferred
Human interferon beta (166 residues), glycosylated, MW=22.5kD. It is produced by mammalian cells (Chinese Hamster Ovary cells) into which the human interferon beta gene has been introduced. The amino acid sequence is identical to that of natural human interferon beta.
Catalog No. 50-208-4232
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.