Learn More
American Research Products Inc Human Interleukin 6 IL 6 ELIS
Supplier:  American Research Products Inc CSBE04638H
Encompass
Human Interleukin 6 IL-6 ELISA KIT from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 2 453 pg/mL Sample serum plasma cell culture supernates tissue homogenates urine Method Sandwich
Catalog No. 50-003-29980