Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IPP (NP_005888, 1 a.a. - 157 a.a.) Partial Recombinant Protein

Catalog No. 89949136 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-136 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-949-136 Supplier Abnova™ Supplier No. P3517
Add to Cart
Add to Cart

Used for SDS-PAGE

The protein encoded by this gene is a member of the kelch family of proteins, which is characterized by a 50 amino acid repeat which interacts with actin. Transcript variants have been described but their full-length nature has not been determined. [provided by RefSeq]

Sequence: MANEDCPKAADSPFSSDKHAQLILAQINKMRNGQHFCDVQLQVGQESFKAHRLVLAASSPYFAALFTGGMKESSKDVVPILGIEAGIFQILLDFIYTGIVNIGVNNVQELIIAADMLQLTEVVHLCCEFLKGQIDPLNCIGIFQFSEQIACHDLLEF

Specifications

Accession Number NP_005888
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 3652
Molecular Weight (g/mol) 17.3kDa
Name IPP (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing Loading 3 ug protein in 15% SDS-PAGE
Quantity 100 μg
Immunogen MANEDCPKAADSPFSSDKHAQLILAQINKMRNGQHFCDVQLQVGQESFKAHRLVLAASSPYFAALFTGGMKESSKDVVPILGIEAGIFQILLDFIYTGIVNIGVNNVQELIIAADMLQLTEVVHLCCEFLKGQIDPLNCIGIFQFSEQIACHDLLEF
Storage Requirements Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KLHL27
Common Name IPP
Gene Symbol IPP
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.