Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IRAK2 Synthetic Peptide

Catalog No. PEP0043
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0043 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0043 Supplier Invitrogen™ Supplier No. PEP0043
Add to Cart
Add to Cart

Synthetic Peptide

IRAK2 Synthetic Peptide, PEP-0043, from Invitrogen.

IRAK2 (interleukin-1 receptor-associated kinase-like 2) binds to the IL-1 type I receptor following IL-1 engagment and triggers intracellular signaling cascades leading to transcriptional up-regulation and mRNA stabilization. The pro-inflammatory cytokine IL-1 induces cellular response through two subunits of its receptor, IL-1 receptor I (IL-1RI) and IL-1 receptor accessory protein (IL-1RAcP). IL-1 receptor-associated kinase (IRAK) mediates activation of NF-kappa-B, which is a pivotal transcription factor mediating inflammatory and immune response. A novel member in the IRAK/Pelle family was recently identified and designated IRAK2. Both IRAK and IRAK2 recruit to the subunits of the IL-1R complex after IL-1 binding and lead to NF-kappa-B activation. IRAKs also associate with Toll-like receptor (TLR) and the dominant negative mutants of IRAKs inhibit LPS-induced NF-kappa-B activation. Members in IRAK/Pelle family play a central role in IL-1R and TLR mediated inflammatory responses to cytokine IL-1 and LPS. IRAK2 is expressed in a variety of human tissues.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 3656
Species Human
Purification Method purified
Accession Number O43187
Common Name Human IRAK2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol IRAK2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.