Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Irisin/FNDC5 DuoSet ELISA
SDP

Catalog No. DY942005 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY942005 1 Kit (5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY942005 Supplier R&D Systems™ Supplier No. DY942005
Add to Cart
Add to Cart
This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human Irisin/FNDC5 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Irisin/FNDC5.

Specifications

Assay Range 250.00 - 8000 pg/mL
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 252995
Host Species Human
Quantity 1 Kit (5 Plates)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target Irisin/FNDC5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.