Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IRX3 Full-length ORF (AAH23667.1, 1 a.a. - 501 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949222 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-222 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-949-222 Supplier Abnova™ Supplier No. H00079191P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

IRX3 is a member of the Iroquois homeobox gene family (see IRX1; MIM 606197) and plays a role in an early step of neural development (Bellefroid et al., 1998 [PubMed 9427753]). Members of this family appear to play multiple roles during pattern formation of vertebrate embryos (Lewis et al., 1999 [PubMed 10370142]).[supplied by OMIM]

Sequence: MSFPQLGYQYIRPLYPSERPGAAGGSGGSAGARGGLGAGASELNASGSLSNVLSSVYGAPYAAAAAAAAAQGYGAFLPYAAELPIFPQLGAQYELKDSPGVQHPAAAAAFPHPHPAFYPYGQYQFGDPSRPKNATRESTSTLKAWLNEHRKNPYPTKGEKIMLAIITKMTLTQVSTWFANARRRLKKENKMTWAPRSRTDEEGNAYGSEREEEDEEEDEEDGKRELELEEEELGGEEEDTGGEGLADDDEDEEIDLENLDGAATEPELSLAGAARRDGDLGLGPISDSKNSDSEDSSEGLEDRPLPVLSLAPAPPPVAVASPSLPSPPVSLDPCAPAPAPASALQKPKIWSLAETATSPDNPRRSPPGAGGSPPGAAVAPSALQLSPAAAAAAAHRLVSAPLGKFPAWTNRPFPGPPPGPRPHPLSLLGSAPPHLLGLPGAAGHPAAAAAFARPAEPEGGTDRCSALEVEKKLLKTAFQPVPRRPQNHLDAALVLSALSSS

Specifications

Accession Number AAH23667.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 79191
Molecular Weight (g/mol) 78.5kDa
Name IRX3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MSFPQLGYQYIRPLYPSERPGAAGGSGGSAGARGGLGAGASELNASGSLSNVLSSVYGAPYAAAAAAAAAQGYGAFLPYAAELPIFPQLGAQYELKDSPGVQHPAAAAAFPHPHPAFYPYGQYQFGDPSRPKNATRESTSTLKAWLNEHRKNPYPTKGEKIMLAIITKMTLTQVSTWFANARRRLKKENKMTWAPRSRTDEEGNAYGSEREEEDEEEDEEDGKRELELEEEELGGEEEDTGGEGLADDDEDEEIDLENLDGAATEPELSLAGAARRDGDLGLGPISDSKNSDSEDSSEGLEDRPLPVLSLAPAPPPVAVASPSLPSPPVSLDPCAPAPAPASALQKPKIWSLAETATSPDNPRRSPPGAGGSPPGAAVAPSALQLSPAAAAAAAHRLVSAPLGKFPAWTNRPFPGPPPGPRPHPLSLLGSAPPHLLGLPGAAGHPAAAAAFARPAEPEGGTDRCSALEVEKKLLKTAFQPVPRRPQNHLDAALVLSALSSS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias IRX-1
Common Name IRX3
Gene Symbol IRX3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.