Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ITGA11 Partial ORF (NP_001004439, 793 a.a. - 893 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949258 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-258 10 μg
89-949-259 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-949-258 Supplier Abnova™ Supplier No. H00022801Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein contains an I domain, is expressed in muscle tissue, dimerizes with beta 1 integrin in vitro, and appears to bind collagen in this form. Therefore, the protein may be involved in attaching muscle tissue to the extracellular matrix. Alternative transcriptional splice variants have been found for this gene, but their biological validity is not determined. [provided by RefSeq]

Sequence: LDARSDLPTAMEYCQRVLRKPAQDCSAYTLSFDTTVFIIESTRQRVAVEATLENRGENAYSTVLNISQSANLQFASLIQKEDSDGSIECVNEERRLQKQVC

Specifications

Accession Number NP_001004439
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 22801
Molecular Weight (g/mol) 36.85kDa
Name ITGA11 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen LDARSDLPTAMEYCQRVLRKPAQDCSAYTLSFDTTVFIIESTRQRVAVEATLENRGENAYSTVLNISQSANLQFASLIQKEDSDGSIECVNEERRLQKQVC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HsT18964
Common Name ITGA11
Gene Symbol ITGA11
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.