Learn More
ACROBiosystems Recombinant Protein;Human Integrin alpha V beta 1 (ITGAV&ITGB1) Heterodimer Protein;HEK293;100UG;IT1,Integrin alpha V beta 1,ITGAV&ITGB1,Integrin,ITGAV,ITGA,ITGB1,ITGB,ITGAVB1
Supplier:  ACROBiosystems IT1H52E1100UG
Encompass_Preferred
Recombinant Protein;Human Integrin alpha V beta 1 (ITGAV&ITGB1) Heterodimer Protein;100UG;Human ITGAV & ITGB1 Heterodimer Protein (IT1-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gln 21 - Asp 728 (ITGB1) (Accession # NP_002201 (ITGAV)& P05556 (ITGB1)).;Human ITGAV & ITGB1 Heterodimer Protein, produced by co-expression of ITGAV and ITGB1, has a calculated MW of 113 kDa (ITGAV) and 83.7 kDa (ITGB1). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 31 (ITGAV) & Gln 21 (ITGB1). The reducing (R) protein migrates as 145 kDa (ITGAV) and 116 kDa (ITGB1) respectively due to glycosylation.;Immobilized Fibronectin at 10 μg/mL (100 μL/well) can bind Human ITGAV & ITGB1 Heterodimer Protein (Cat. No. IT1-H52E1) with a linear range of 0.156-1.25 μg/mL (QC tested).
Catalog No. 50-108-9641
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.