Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ITIH1 Synthetic Peptide

Catalog No. PEP301
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP301 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP301 Supplier Invitrogen™ Supplier No. PEP301
Add to Cart
Add to Cart

Synthetic Peptide

ITI-HC1 Synthetic Peptide, PEP-301, from Invitrogen™.

The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]).
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 3697
Species Human
Purification Method purified
Accession Number P19827
Common Name Human ITIH1
Concentration 1 mg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Lyophilized
Formulation water with no preservative
For Use With (Application) Blocking Assay, Control
Gene Alias CYCSP39, cytochrome c, somatic pseudogene 39, HCP39
Gene Symbol ITIH1
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.