Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ITIH5 (NP_085046.5) VersaClone cDNA
SDP

Catalog No. RDC3317 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC3317 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC3317 Supplier R&D Systems™ Supplier No. RDC3317
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Gene ITIH5
Accession Number NP_085046.5
Gene ID (Entrez) 80760
Gene Alias inter-alpha (globulin) inhibitor H5, inter-alpha-inhibitor heavy chain 5, inter-alpha-trypsin inhibitor heavy chain H5, ITI heavy chain H5, ITIH5 inter-alpha-trypsin inhibitor heavy chain family, member 5, ITI-HC5, KIAA1953, MGC10848, PP14776
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.