Learn More
Abcam Human Jagged1 Peptide, Synthetic, Blocking, 1 mg
Description
- Protein Name: Jagged1
- Type: Synthetic
- Purity: >70%
- Tested Applications: BL
- Species Reactivity: Human
- Formulation: Liquid, pH 6.75
- Storage Buffer Constituents: 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Peptide-based Substrate |
| Shipping Condition | Blue Ice |
| Target Molecule | Jagged1 (Human) |
| Reactivity | Human |
| Label or Dye | Unconjugated |
| Form | Liquid |
| Product Type | Blocker and Diluent |
| Content And Storage | Store at -20°C. |
| Buffer | 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH6.75 |
| Concentration | 1 mg/mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.