Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Jarid1A/KDM5A Antibody, R&D Systems™

Catalog No. MAB11705100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11705100 100 μg
MAB11705SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB11705100 Supplier R&D Systems Supplier No. MAB11705100
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Jarid1A Monoclonal antibody specifically detects Jarid1A in Human samples. It is validated for Western Blot,Simple Western

Specifications

Antigen Jarid1A
Applications Western Blot, Simple Western
Classification Monoclonal
Clone 1102542
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Simple Western 2-10 μg/mL
Formulation Lyophilized from a 0.2 Âμm filtered solution in PBS with Trehalose.
Gene Accession No. P29375
Gene Alias EC 1.14.11, EC 1.14.11.-, EC 1.14.11.67, Histone demethylase JARID1A, JARID1AJumonji, AT rich interactive domain 1A (RBP2-like), Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji/ARID domain-containing protein 1A, KDM5A, lysine (K)-specific demethylase 5A, RBBP2, RBBP-2, RBBP2lysine-specific demethylase 5A, RBP2, RBP2retinoblastoma binding protein 2, Retinoblastoma-binding protein 2jumonji, AT rich interactive domain 1A
Host Species Mouse
Immunogen E. coli -derived recombinant human Jarid1A/KDM5A, Lys127-Ser287, Accession # P29375
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5927
Reconstitution Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.