Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Kallikrein 1 DuoSet ELISA
SDP

Catalog No. DY233705 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY233705 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY233705 Supplier R&D Systems™ Supplier No. DY233705
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)

For the development of sandwich ELISAs to measure natural and recombinant human Kallikrein 1

The Human Kallikrein 1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Kallikrein 1.

Specifications

Assay Range 46.90 to 3000 pg/mL
Detectable Analytes Kallikrein 1
Assay Solid Phase ELISA
Conjugate Biotin
Cross Reactivity <50%cross-species reactivity observed with species tested
Immunoassay Kit Format 96-well Strip Plate
Product Type ELISA Kit
Sample Type Cell Culture Supernatant
Sufficient For To run ELISAs on at least 5 x 96-well plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3816, 16612
Host Species Human
Quantity 5 Plates
Sample Volume 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.