Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Kallikrein 15 Fc Chimera Recombinant Protein

Catalog No. PIRP88083
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP88083 Supplier Invitrogen™ Supplier No. RP88083
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (660 μL) be added to the vial to prepare a stock solution of 0.151515151515152 mg/mL. Concentration is measured by UV.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H2R5
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 55554
Molecular Weight (g/mol) 53.1 kDa
Name Human Kallikrein 15 Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias ACO; ACO protease; HSRNASPH; kallikrein 15; kallikrein related peptidase 15; kallikrein related-peptidase 15; kallikrein-15; kallikrein-like serine protease; kallikrein-related peptidase 15; KLK15; KLK-15; prostinogen
Common Name Kallikrein 15
Gene Symbol KLK15
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human KLK15, amino acids Met1-Asn256 (Accession # NP_059979.2) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.