Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KCNJ4 Full-length ORF (CAK54537.1, 1 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949546 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-546 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-949-546 Supplier Abnova™ Supplier No. H00003761P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Several different potassium channels are known to be involved with electrical signaling in the nervous system. One class is activated by depolarization whereas a second class is not. The latter are referred to as inwardly rectifying K+ channels, and they have a greater tendency to allow potassium to flow into the cell rather than out of it. This asymmetry in potassium ion conductance plays a key role in the excitability of muscle cells and neurons. The protein encoded by this gene is an integral membrane protein and member of the inward rectifier potassium channel family. The encoded protein has a small unitary conductance compared to other members of this protein family. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MHGHSRNGQAHVPRRKRRNRFVKKNGQCNVYFANLSNKSQRYMADIFTTCVDTRWRYMLMIFSAAFLVSWLFFGLLFWCIAFFHGDLEASPGVPAAGGPAAGGGGAAPVAPKPCIMHVNGFLGAFLFSVETQTTIGYGFRCVTEECPLAVIAVVVQSIVGCVIDSFMIGTIMAKMARPKKRAQTLLFSHHAVISVRDGKLCLMWRVGNLRKSHIVEAHVRAQLIKPYMTQEGEYLPLDQRDLNVGYDIGLDRIFLVSPIIIVHEIDEDSPLYGMGKEELESEDFEIVVILEGMVEATAMTTQARSSYLASEILWGHRFEPVVFEEKSHYKVDYSRFHKTYEVAGTPCCSARELQESKITVLPAPPPPPSAFCYENELALMSQEEEEMEEEAAAAAAVAAGLGLEAGSKEEAGIIRMLEFGSHLDLERMQASLPLDNISYRRESAI

Specifications

Accession Number CAK54537.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3761
Molecular Weight (g/mol) 75.35kDa
Name KCNJ4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MHGHSRNGQAHVPRRKRRNRFVKKNGQCNVYFANLSNKSQRYMADIFTTCVDTRWRYMLMIFSAAFLVSWLFFGLLFWCIAFFHGDLEASPGVPAAGGPAAGGGGAAPVAPKPCIMHVNGFLGAFLFSVETQTTIGYGFRCVTEECPLAVIAVVVQSIVGCVIDSFMIGTIMAKMARPKKRAQTLLFSHHAVISVRDGKLCLMWRVGNLRKSHIVEAHVRAQLIKPYMTQEGEYLPLDQRDLNVGYDIGLDRIFLVSPIIIVHEIDEDSPLYGMGKEELESEDFEIVVILEGMVEATAMTTQARSSYLASEILWGHRFEPVVFEEKSHYKVDYSRFHKTYEVAGTPCCSARELQESKITVLPAPPPPPSAFCYENELALMSQEEEEMEEEAAAAAAVAAGLGLEAGSKEEAGIIRMLEFGSHLDLERMQASLPLDNISYRRESAI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HIR/HIRK2/HRK1/IRK3/Kir2.3/MGC142066/MGC142068
Common Name KCNJ4
Gene Symbol KCNJ4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.