Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KCTD7 Full-length ORF (NP_694578.1, 1 a.a. - 289 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949604 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-604 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-949-604 Supplier Abnova™ Supplier No. H00154881P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The KCTD gene family, including KCTD7, encode predicted proteins that contain N-terminal domain that is homologous to the T1 domain in voltage-gated potassium channels (see KCNA1; MIM 176260). KCTD7 displays a primary sequence and hydropathy profile indicating intracytoplasmic localization. EST database analysis showed that KCTD7 is expressed in human and mouse brain (Van Bogaert et al., 2007 [PubMed 17455289]).[supplied by OMIM]

Sequence: MVVVTGREPDSRRQDGAMSSSDAEDDFLEPATPTATQAGHALPLLPQEFPEVVPLNIGGAHFTTRLSTLRCYEDTMLAAMFSGRHYIPTDSEGRYFIDRDGTHFGDVLNFLRSGDLPPRERVRAVYKEAQYYAIGPLLEQLENMQPLKGEKVRQAFLGLMPYYKDHLERIVEIARLRAVQRKARFAKLKVCVFKEEMPITPYECPLLNSLRFERSESDGQLFEHHCEVDVSFGPWEAVADVYDLLHCLVTDLSAQGLTVDHQCIGVCDKHLVNHYYCKRPIYEFKITWW

Specifications

Accession Number NP_694578.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 154881
Molecular Weight (g/mol) 59.5kDa
Name KCTD7 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MVVVTGREPDSRRQDGAMSSSDAEDDFLEPATPTATQAGHALPLLPQEFPEVVPLNIGGAHFTTRLSTLRCYEDTMLAAMFSGRHYIPTDSEGRYFIDRDGTHFGDVLNFLRSGDLPPRERVRAVYKEAQYYAIGPLLEQLENMQPLKGEKVRQAFLGLMPYYKDHLERIVEIARLRAVQRKARFAKLKVCVFKEEMPITPYECPLLNSLRFERSESDGQLFEHHCEVDVSFGPWEAVADVYDLLHCLVTDLSAQGLTVDHQCIGVCDKHLVNHYYCKRPIYEFKITWW
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EPM3/FLJ32069
Common Name KCTD7
Gene Symbol KCTD7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.