Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KDR (NM_002253, 789 a.a. - end) Partial Recombinant Protein with GST-tag expressed in Sf9 cells

Catalog No. 89949617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-617 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-949-617 Supplier Abnova™ Supplier No. P3871

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for Func, SDS-PAGE

Vascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc.. Mutations of this gene are implicated in infantile capillary hemangiomas. [provided by RefSeq]

Specifications

Accession Number NM_002253
For Use With (Application) Functional Study, SDS-PAGE
Formulation In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol)
Gene ID (Entrez) 3791
Molecular Weight (g/mol) 110kDa
Name KDR (Human) Recombinant Protein
Preparation Method Insect cell (Sf9) expression system
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CD309/FLK1/VEGFR/VEGFR2
Common Name KDR
Gene Symbol KDR
Biological Activity The specific activity was determined to be 58nmol /min/mg as per Activity Assay Protocol.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Insect cell (Sf9) expression system
Purity or Quality Grade >70% by densitometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.