Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Kell Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7064712
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1914SP 25 μg
MAB1914 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1914SP Supplier R&D Systems Supplier No. MAB1914SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kell Monoclonal specifically detects Kell in Human samples. It is validated for Western Blot.

Specifications

Antigen Kell
Applications Western Blot
Classification Monoclonal
Clone 195031
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P23276
Gene Alias CD238, CD238 antigen, EC 3.4.24.-, ECE3, KEL, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase
Gene Symbols KEL
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kell Asn68-Trp732 Accession # P23276
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3792
Test Specificity Detects human Kell in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human ECE-2, recombinant human Neprilysin, or recombinant mouse Kell is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.