Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KGF/FGF-7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067889
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB251SP 25 μg
MAB251100 100 μg
MAB251500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB251SP Supplier R&D Systems Supplier No. MAB251SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

KGF/FGF-7 Monoclonal specifically detects KGF/FGF-7 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen KGF/FGF-7
Applications Neutralization, ELISA
Classification Monoclonal
Clone 29522
Conjugate Unconjugated
Dilution Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P21781
Gene Alias FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Gene Symbols FGF7
Host Species Mouse
Immunogen E. coli-derived recombinant human KGF/FGF-7 Cys32-Thr194 Accession # P21781
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2252
Test Specificity Detects human KGF/FGF-7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGF-3, rhFGF-4, rhFGF-5, rhFGF-6, rhFGF acidic, or rhFGF basic is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.