Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Ki67/MKI67 DuoSet ELISA
SDP

Catalog No. DY761705 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
Five 96-well plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DY761705 Five 96-well plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY761705 Supplier R&D Systems™ Supplier No. DY761705
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human Ki-67.

The Human Ki-67/MKI67 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Ki-67/MKI67.

Specifications

Assay Range 6.25 to 400pg/mL
Conjugate Biotin
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
Sufficient For Five 96-well plates
For Use With (Application) ELISA
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, MKI67, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105, TSG126
Target Ki67/MKI67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.